Tantal ≥99,9% (Metall-Basis), dünne Folie, Dicke 0.0025 mm (0.0001 in)
Lieferant: Thermo Scientific
40716.FF
40716.FI
40716.GH
40716.FFEA
346
EUR
40716.FF
40716.FI
40716.GH
Tantal ≥99,9% (Metall-Basis), dünne Folie, Dicke 0.0025 mm (0.0001 in)
Tantalum
Purity: 99.9%
CAS No.: 7440-25-7
Physical Form: ≈0.026g/25x25mm, not certified pinhole free
Odour: Odourless
MDL No.: MFCD00011252
Formel:
Ta Molecular Weight: 180,95 g/mol Siedepunkt: 5427 °C (1013 hPa) Schmelzpunkt: 2996 °C Dichte: 16,69 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011252 CAS-Nummer: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...