Clindamycin hydrochloride, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Cleocin Hydrochloride, (2S,4R)-N-((1S,2S)-2-Chloro-1-((2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(methylthio)tetrahydro-2H-pyran-2-yl)propyl)-1-methyl-4-propylpyrrolidine-2-carboxamide hydrochloride, 7[S]-Chloro-7-deoxylincomycin, Clindamycin HCl

M-Clarity™ Qualitätsstufe = MQ200

C5269-50MG C5269-100MG C5269-10MG
SIALC5269-50MGEA 503 EUR
SIALC5269-50MG SIALC5269-100MG SIALC5269-10MG
Clindamycin hydrochloride, Sigma-Aldrich®
Clindamycin hydrochloride

Clindamycin is a semi-synthetic, lincosamide antibiotic that is prepared from lincomycin. It inhibits bacterial protein synthesis by hydrogen bond interactions with the 23S rRNA component of the 50S ribosomal subunit thus inducing dissociation of the peptidyl-t-RNA complex. It has antibacterial activity against Gram-positive cocci and antiprotozoal activity against taxoplasma.


  • Effective against anaerobic species
  • Mode of action: protein synthesis
  • Antibiotic activity spectrum: gram-negative bacteria, gram-positive bacteria


Clindamycin is used to study bacterial infections, such as group B streptococcal disease, bacterial resistance and plasma protein binding.


Crystalline


Soluble in water (50 mg/ml)

Formel: C₁₈H₃₃ClN₂O₅S·HCl
Molecular Weight: 461.45 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07793327
CAS-Nummer: 21462-39-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR