Palladium ≥99,8% (Metall-Basis), Granulat
Lieferant: Thermo Fisher Scientific
12237.03 12237.06 12237.14
12237.03EA 331 EUR
12237.03 12237.06 12237.14
Palladium ≥99,8% (Metall-Basis), Granulat
Palladium
Purity: 99.8% CAS No.: 7440-05-3 Odour: Odourless MDL No.: MFCD00011167
Formel: Pd
Molecular Weight: 106.420 g/mol
Dichte: 12.02 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011167
CAS-Nummer: 7440-05-3
EINECS: 231-115-6