Molekularsieb 3A (0,3 nm, 3 Å) with moisture indicator ~10 mesh, Ø ~2 mm, Supelco®
Lieferant: Merck
Synonyme: Molekularsieb A3
M-Clarity™ Qualitätsstufe = MQ200
1.05734.0250 1.05734.1000
1.05734.0250EA 96.5 EUR
1.05734.0250 1.05734.1000
Molekularsieb 3A (0,3 nm, 3 Å) with moisture indicator ~10 mesh, Ø ~2 mm, Supelco®
Molekularsieb 3A (0,3 nm, 3 Å)
Formel: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL: MFCD00131613
CAS-Nummer: 308080-99-1