4-Chlorchalkon

Lieferant: MENAI

Synonyme: 4-Chlorbenzylidenacetophenon

IP23.100 IP23.250
MENAIP23.100EA 0 EUR
MENAIP23.100 MENAIP23.250
4-Chlorchalkon
4-Chlorchalkon
Formel: C₁₅H₁₁ClO
Molecular Weight: 242.7 g/mol
MDL: MFCD00016345
CAS-Nummer: 956-04-7
EINECS: 213-476-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR