2-Nitrochalkon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
A17763.06
A17763.14
A17763.06EA
43.5
EUR
A17763.06
A17763.14
2-Nitrochalkon ≥97%
2-Nitrochalkon
Purity: 97%
CAS No.: 7473-93-0
Molecular Formula: C15H11NO3
Formula Weight: 253.26
Melting Point: 124-128°
MDL No.: MFCD00031069
Formel:
C₁₅H₁₁NO₃ Molecular Weight: 253,26 g/mol Schmelzpunkt: 124…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00031069 CAS-Nummer: 7473-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...