4-Methoxychalkon ≥97%

Lieferant: Thermo Scientific

Synonyme: 4-Methoxybenzylidenacetophenon

L03319.06 L03319.14
L03319.06EA 20.2 EUR
L03319.06 L03319.14
4-Methoxychalkon ≥97%
4-Methoxychalkon

Purity: 97% CAS No.: 959-33-1 Molecular Formula: C16H14O2 Formula Weight: 238.29 Melting Point: 73-76° Beilstein Reference: 978742

Formel: C₁₆H₁₄O₂
Molecular Weight: 238,29 g/mol
Siedepunkt: 187 °C (19 mmHg)
Schmelzpunkt: 73…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017179
CAS-Nummer: 959-33-1
EINECS: 213-499-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR