4'-Methoxychalkon ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4'-Methoxybenzylidenacetophenon, α-Styryl-p-anisylketon

B22158.06 B22158.14 B22158.22
B22158.06EA 26.3 EUR
B22158.06 B22158.14 B22158.22
4'-Methoxychalkon ≥97%
4'-Methoxychalkon

Purity: 97% CAS No.: 959-23-9 Molecular Formula: C16H14O2 Formula Weight: 238.29 Melting Point: 101-103° Beilstein Reference: 644640

Formel: C₁₆H₁₄O₂
Molecular Weight: 238,29 g/mol
Schmelzpunkt: 101…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008407
CAS-Nummer: 959-23-9
EINECS: 213-495-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR