4'-Hydroxychalkon
Lieferant: FLUOROCHEM
Synonyme:
4'-Hydroxybenzylidenacetophenon
003112-5G
003112-25G
FLUO003112-5GEA
36.9
EUR
FLUO003112-5G
FLUO003112-25G
4'-Hydroxychalkon
4'-Hydroxychalkon
Formel:
C₁₅H₁₂O₂ Molecular Weight: 224.26 g/mol |
MDL:
MFCD00016484 CAS-Nummer: 2657-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...