Molekularsieb 3A (0,3 nm, 3 Å), Perlen 2 mm, Supelco®
Lieferant: Merck
Synonyme:
Molekularsieb A3
M-Clarity™ Qualitätsstufe = MQ200
1.05704.0250
1.05704.1000
1.05704.9010
1.05704.0250EA
96.5
EUR
1.05704.0250
1.05704.1000
1.05704.9010
Molekularsieb 3A (0,3 nm, 3 Å), Perlen 2 mm, Supelco®
Molekularsieb 3A (0,3 nm, 3 Å)
Formel:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00131613 CAS-Nummer: 308080-99-1 EINECS: 215-283-8 |
Spezifikation Testergebnisse
Solubility | (20 °C) insoluble |
Bulk Density | 700 - 750 kg/m3 |
Synonyms | Sodium aluminium silicate |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...