Molekularsieb A4 (0,4 nm, 4 Å), Plätzchen, Ø 1 - 2mm (0.04-0.08in)
Lieferant: Thermo Scientific
Synonyme:
Molecular sieve 0.4 nm, Molecular sieves pack 4A, Linde 4A, Sodium aluminium silicate
87956.A1
87956.A7
87956.A1EA
129
EUR
87956.A1
87956.A7
Molekularsieb A4 (0,4 nm, 4 Å), Plätzchen, Ø 1 - 2mm (0.04-0.08in)
Molekularsieb A4 (0,4 nm, 4 Å)
Dichte:
1,1…2.2 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03457537 CAS-Nummer: 70955-01-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...