Bromoform-[D1], Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tribrommethan-D1
M-Clarity™ Qualitätsstufe = MQ200
329320-10G
329320-25G
SIAL329320-10GEA
185
EUR
SIAL329320-10G
SIAL329320-25G
Bromoform-[D1], Sigma-Aldrich®
Bromoform-[D1]
Formel:
Br₃CD Molecular Weight: 253,72 g/mol |
CAS-Nummer:
2909-52-6 EINECS: 220-823-0 UN: 2515 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...