2-Chlor-2-methylpropan-[D9], Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butylchlorid-D9

M-Clarity™ Qualitätsstufe = MQ200

185949-1G 185949-5G
SIAL185949-1GEA 262 EUR
SIAL185949-1G SIAL185949-5G
2-Chlor-2-methylpropan-[D9], Sigma-Aldrich®
2-Chlor-2-methylpropan-[D9]
Formel: D₃CCCl(CD₃)CD₃
Molecular Weight: 101.5 g/mol
CAS-Nummer: 918-20-7
EINECS: 213-041-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR