Benzoesäure-[Ring-D5] (98% D)

Lieferant: LGC Standards PROMOCHEM

Synonyme: 2,3,4,5,6-Pentadeuterobenzoesäure

CIL-DLM-122-1 CIL-DLM-122-5
PROOCIL-DLM-122-1EA 137 EUR
PROOCIL-DLM-122-1 PROOCIL-DLM-122-5
Benzoesäure-[Ring-D5] (98% D)
Benzoesäure-[Ring-D5]
Formel: C₆D₅COOH
Molecular Weight: 127,08 g/mol
Schmelzpunkt: 120…122 °C
CAS-Nummer: 1079-02-3
EINECS: 214-089-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR