Chrysen-[D12] (98% D)

Lieferant: LGC Standards PROMOCHEM

CILDLM-261-0.1 CIL-DLM-261-1
PROOCILDLM-261-0.1EA 217 EUR
PROOCILDLM-261-0.1 PROOCIL-DLM-261-1
Chrysen-[D12] (98% D)
Chrysen-[D12]
Formel: C₁₈D₁₂
CAS-Nummer: 1719-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR