Chrysen-[D12], Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

364614-250MG 364614-50MG
SIAL364614-250MGEA 433 EUR
SIAL364614-250MG SIAL364614-50MG
Chrysen-[D12], Sigma-Aldrich®
Chrysen-[D12]
Formel: C₁₈D₁₂
CAS-Nummer: 1719-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR