Fulleren C70 97%, Pulver

Lieferant: Thermo Scientific
Achtung

Synonyme: Buckminster-Fulleren C70

39720.MA 39720.MF 39720.MD
39720.MAEA 114 EUR
39720.MA 39720.MF 39720.MD
Fulleren C70 97%, Pulver
Fulleren C70

Purity: 97% CAS No.: 115383-22-7 Density: 1.7 Appearance: Black Boiling Point: 500-600° subl. UN No.: 1325 MDL No.: MFCD00282904

Formel: C₇₀
Molecular Weight: 840,77 g/mol
Siedepunkt: 500…600 °C (Subl.)
Schmelzpunkt: > 280 °C
Dichte: 1,7
Lagertemperatur: Raumtemperatur
MDL: MFCD00282904
CAS-Nummer: 115383-22-7
UN: 1325
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR