trans-Platin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-Diammindichloridoplatin(II)

P1525-250MG P1525-1G
SIALP1525-250MGEA 97.1 EUR
SIALP1525-250MG SIALP1525-1G
trans-Platin, Sigma-Aldrich®
trans-Platin
Formel: H₆Cl₂N₂Pt
Molecular Weight: 300,05 g/mol
Schmelzpunkt: 340 °C
Dichte: 3,746 g/cm³ (20 °C)
MDL: MFCD00011623
CAS-Nummer: 14913-33-8
EINECS: 238-980-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR