D(+)-Galactose, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2R,3S,4S,5R)-Pentahydroxyhexanal
MQ300
G0750-10G
G0750-100G
G0750-10MG
G0750-1KG
G0750-25G
G0750-500G
G0750-50G
SIALG0750-10GEA
35
EUR
SIALG0750-10G
SIALG0750-100G
SIALG0750-10MG
SIALG0750-1KG
SIALG0750-25G
SIALG0750-500G
SIALG0750-50G
D(+)-Galactose, Sigma-Aldrich®
D(+)-Galactose
Formel:
C₆H₁₂O₆ Molecular Weight: 180.16 g/mol Siedepunkt: 527 Schmelzpunkt: 163…169 °C Dichte: 1.5 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151230 CAS-Nummer: 59-23-4 EINECS: 200-416-4 Merck Index: 13,04356 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...