D(+)-Galactose, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(2R,3S,4S,5R)-Pentahydroxyhexanal
MQ200
G6404-100G
G6404-25G
SIALG6404-100GEA
414
EUR
SIALG6404-100G
SIALG6404-25G
D(+)-Galactose, Sigma-Aldrich®
D(+)-Galactose
Formel:
C₆H₁₂O₆ Molecular Weight: 180.16 g/mol Siedepunkt: 527 Schmelzpunkt: 163…169 °C Dichte: 1.5 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151230 CAS-Nummer: 59-23-4 EINECS: 200-416-4 Merck Index: 13,04356 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...