L(-)-Galactose, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2S,3R,4R,5S)-Pentahydroxyhexanal

M-Clarity™ Qualitätsstufe = MQ100

G7134-100MG G7134-1G G7134-250MG G7134-25MG
SIALG7134-100MGEA 317 EUR
SIALG7134-100MG SIALG7134-1G SIALG7134-250MG SIALG7134-25MG
L(-)-Galactose, Sigma-Aldrich®
L(-)-Galactose
Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063833
CAS-Nummer: 15572-79-9
EINECS: 239-630-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR