L(-)-Mannose 99+%

Lieferant: Thermo Scientific

Synonyme: (2R,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal, L-(-)-Mannose

227590010 227592500 227590050
ACRO227590010EA 129 EUR
ACRO227590010 ACRO227592500 ACRO227590050
L(-)-Mannose 99+%
L(-)-Mannose
Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
Siedepunkt: 376 °C (1013 hPa)
Schmelzpunkt: 129…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00136021
CAS-Nummer: 10030-80-5
EINECS: 233-080-2

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -13.5° to -15.5° (20°C, 589 nm) (c=4, water, NH3)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR