L(-)-Xylose 99+%

Lieferant: Thermo Scientific

Synonyme: (2S,3R,4S)-2,3,4,5-Tetrahydroxypentanal, L-(-)-Xylose

225990050 225990250 225991000
ACRO225990050EA 47.9 EUR
ACRO225990050 ACRO225990250 ACRO225991000
L(-)-Xylose 99+%
L(-)-Xylose
Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Siedepunkt: 331 °C (1013 hPa)
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151096
CAS-Nummer: 609-06-3
EINECS: 210-174-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥99.0 % (ELSD)
Specific optical rotation -18° to -20° (20°C, 589 nm) (c=10, H2O) (after 24 h)
Loss on ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR