L(-)-Xylose ≥99%

Lieferant: Thermo Scientific

Synonyme: (2S,3R,4S)-2,3,4,5-Tetrahydroxypentanal, L-(-)-Xylose

B21622.06 B21622.14 B21622.22
B21622.06EA 44.9 EUR
B21622.06 B21622.14 B21622.22
L(-)-Xylose ≥99%
L(-)-Xylose

Purity: 99% CAS No.: 609-06-3 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 147-151° Beilstein Reference: 1723080

Formel: C₅H₁₀O₅
Molecular Weight: 150,13 g/mol
Siedepunkt: 331 °C (1013 hPa)
Schmelzpunkt: 147…151 °C
Dichte: 1,525
Lagertemperatur: Raumtemperatur
MDL: MFCD00151096
CAS-Nummer: 609-06-3
EINECS: 210-174-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR