Agarose

Lieferant: Serva

11381.01 11381.03
SERA11381.01EA 0 EUR
SERA11381.01 SERA11381.03
Agarose
Agarose
Schmelzpunkt: 60…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081294
CAS-Nummer: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR