Stärke (aus Kartoffel), Pulver, reinst
Lieferant: Thermo Scientific
419695000
419690025
ACRO419695000EA
44.2
EUR
ACRO419695000
ACRO419690025
Stärke (aus Kartoffel), Pulver, reinst
Starch
Formel:
(C₆H₁₀O₅)n Molecular Weight: 162,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082026 CAS-Nummer: 9005-25-8 EINECS: 232-679-6 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | 18.0 % to 20.5 % (1 g, 130°C, 90 min) |
Ash | ≤0.5 % (900°C) |
Microbial contamination | (Salmonella) Absent in 25 g |
pH | 6 to 8 (50 % suspension in water) |
Sulfite (as SO2) | ≤10 ppm |
Arsenic (As) | ≤0.07 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤0.1 ppm |
Mercury (Hg) | ≤0.05 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...