Acrylamid-Bisacrylamid (29:1) 30% aqueous solution
Lieferant: Serva
Gefahr
Synonyme: Acrylamid-Bis
10687.01 10687.02 10687.03
SERA10687.01EA 60.4 EUR
SERA10687.01 SERA10687.02 SERA10687.03
Acrylamid-Bisacrylamid (29:1) 30% aqueous solution
Acrylamid-Bis (32:1)
MDL: MFCD00080848
UN: 3426
ADR: 6.1 III