Acrylamid-Bisacrylamid (29:1) 40% aqueous solution
Lieferant: Serva
Gefahr
Synonyme: Acrylamid-Bis
10680.01 10680.02 10680.03
SERA10680.01EA 65.9 EUR
SERA10680.01 SERA10680.02 SERA10680.03
Acrylamid-Bisacrylamid (29:1) 40% aqueous solution
Acrylamid-Bis (32:1)
Lagertemperatur: Raumtemperatur
MDL: MFCD00080848
UN: 3426
ADR: 6.1 III