Acrylamid-Bisacrylamid (37.5:1) 30% aqueous solution
Lieferant: PanReac AppliChem
Gefahr
Synonyme: Acrylamid-Bis
A3626.0500 A3626.1000
A3626.0500EA 81.1 EUR
A3626.0500 A3626.1000
Acrylamid-Bisacrylamid (37.5:1) 30% aqueous solution
Acrylamid-Bis (32:1)
Dichte: 1.03 g/cm³ (20 °C)
MDL: MFCD00080848
UN: 3426
ADR: 6.1 III