Acrylamid-Bisacrylamid (37.5:1) 30% in wässriger Lösung
Lieferant: Serva
|
Gefahr
|
Synonyme:
Acrylamid-Bis
10688.01
10688.02
10688.03
SERA10688.01EA
60.4
EUR
SERA10688.01
SERA10688.02
SERA10688.03
Acrylamid-Bisacrylamid (37.5:1) 30% in wässriger Lösung
Acrylamid-Bis (32:1)
Dichte:
1,03 g/cm³ (20 °C) |
MDL:
MFCD00080848 UN: 3426 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...