Acrylamid-Bisacrylamid (37.5:1) 40% aqueous solution, crystallized_4
Lieferant: BIOMOL RESEARCH LABORATORIES
Gefahr
Synonyme: Acrylamid-Bis
54535-1 54535-3
BMOL54535-1EA 0 EUR
BMOL54535-1 BMOL54535-3
Acrylamid-Bisacrylamid (37.5:1) 40% aqueous solution, crystallized_4
Acrylamid-Bis (32:1)
Lagertemperatur: Raumtemperatur
MDL: MFCD00080848
UN: 3426
ADR: 6.1 III