Acrylamid-Bisacrylamid (37.5:1) 40% aqueous solution
Lieferant: Serva
|
Gefahr
|
Synonyme:
Acrylamid-Bis
10681.02
10681.03
SERA10681.02EA
226
EUR
SERA10681.02
SERA10681.03
Acrylamid-Bisacrylamid (37.5:1) 40% aqueous solution
Acrylamid-Bis (32:1)
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00080848 UN: 3426 ADR: 6.1 III |