TRITON™ X-405 (Polyethylenglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]ether, Polyethylenglycol-tert-octylphenylether)
Lieferant: Uptima
Synonyme:
Polyethylenglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]ether, 4-(1,1,3,3-Tetramethylbutyl)phenylpolyethylenglycol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol, Alkylphenylpolyethylenglycol
UP158536
UP158537
UPTIUP158536EA
0
EUR
UPTIUP158536
UPTIUP158537
TRITON™ X-405 (Polyethylenglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]ether, Polyethylenglycol-tert-octylphenylether)
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether)
Formel:
C₂₈H₅₀O₈ Molecular Weight: 514.69 g/mol Siedepunkt: 120 Schmelzpunkt: –4 °C Dichte: 1.05 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01779734 CAS-Nummer: 9002-93-1 Merck Index: 12,06858 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...