Tween® 20 (Polysorbat)

Lieferant: Uptima

Synonyme: Polyoxyethylene sorbitan monolaurate

UP158740 UP158741
UPTIUP158740EA 0 EUR
UPTIUP158740 UPTIUP158741
Tween® 20 (Polysorbat)
Tween® 20 (Polysorbat)
Formel: C₅₈H₁₁₄O₂₆
Molecular Weight: 1227.53 g/mol
Dichte: 1.1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00165986
CAS-Nummer: 9005-64-5
EINECS: 500-018-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR