Tannin ACS
Lieferant: Thermo Fisher Scientific
Synonyme:
Gerbsäure, Tannin
419995000
419990025
ACRO419995000EA
88.4
EUR
ACRO419995000
ACRO419990025
Tannin ACS
Tannin
Formel:
C₇₆H₅₂O₄₆ Molecular Weight: 1701.2 g/mol Schmelzpunkt: 218 °C (Zers.) Flammpunkt: 198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066397 CAS-Nummer: 1401-55-4 EINECS: 215-753-2 Merck Index: 12,09221 |
Spezifikation Testergebnisse
Appearance | Light brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤12.0 % (105°C) |
Identification | Passes test |
Residue after ignition | ≤0.5 % |
Zinc (Zn) |
= |
Heavy metals (as Pb) | ≤30 ppm |
Sugars, dextrin | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...