Tannin

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Gerbsäure, Tannin

A17022.0B A17022.30 A17022.PG
A17022.0BEA 132 EUR
A17022.0B A17022.30 A17022.PG
Tannin
Tannin

CAS No.: 1401-55-4 Molecular Formula: C76H52O46 Formula Weight: 1701.23 Odour: Odourless pH: 3.5 Melting Point: ca 218° dec. Beilstein Reference: 8186396 Merck Reference: 14,9052 MDL No.: MFCD00066397

Formel: C₇₆H₅₂O₄₆
Molecular Weight: 1701.2 g/mol
Schmelzpunkt: ∼ 218 °C (Zers.)
Dichte: 1 g/cm³ (20 °C)
Flammpunkt: 199
Lagertemperatur: Raumtemperatur
MDL: MFCD00066397
CAS-Nummer: 1401-55-4
EINECS: 215-753-2
Merck Index: 12,09221

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR