Polyvinylpyrrolidon, M.W. ∼ 10,000 (K15)

Lieferant: Serva

33422.01 33422.02
SERA33422.01EA 55.2 EUR
SERA33422.01 SERA33422.02
Polyvinylpyrrolidon, M.W. ∼ 10,000 (K15)
Polyvinylpyrrolidon
Formel: (C₆H₉NO)n
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR