Polyvinylpyrrolidon, M.W. ∼ 24,000 (K25)

Lieferant: Serva

33420.02 33420.03
SERA33420.02EA 62.8 EUR
SERA33420.02 SERA33420.03
Polyvinylpyrrolidon, M.W. ∼ 24,000 (K25)
Polyvinylpyrrolidon
Formel: (C₆H₉NO)n
Lagertemperatur: Raumtemperatur
MDL: MFCD01076626
CAS-Nummer: 9003-39-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR