DL-Arginin ≥98%
Lieferant: Thermo Scientific
Synonyme:
(±)-Arginin, α-Amino-δ-guanidinovaleriansäure, (±)-α-Amino-δ-guanidinovaleriansäure, H-Arg-OH, Arginin
A17520.06
A17520.14
A17520.06EA
34
EUR
A17520.06
A17520.14
DL-Arginin ≥98%
DL-Arginin
Purity: 98%
CAS No.: 7200-25-1
Molecular Formula: C6H14N4O2
Formula Weight: 174.20
Melting Point: ca 230° dec.
Beilstein Reference: 1725411
Formel:
C₆H₁₄N₄O₂ Molecular Weight: 174,20 g/mol Schmelzpunkt: ∼ 230 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063117 CAS-Nummer: 7200-25-1 EINECS: 230-571-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...