DL-Arginin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-Arginin, α-Amino-δ-guanidinovaleriansäure, (±)-α-Amino-δ-guanidinovaleriansäure, H-Arg-OH, Arginin

11020-25G 11020-5G
SIAL11020-25GEA 91.8 EUR
SIAL11020-25G SIAL11020-5G
DL-Arginin, Sigma-Aldrich®
DL-Arginin
Formel: C₆H₁₄N₄O₂
Molecular Weight: 174,20 g/mol
MDL: MFCD00063117
CAS-Nummer: 7200-25-1
EINECS: 230-571-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR