DL-Methionin

Lieferant: BACHEM BIOCHEMICA

Synonyme: Methioninum racemicum, Methionin racemisch, H-DL-Met-OH, (±)-2-Amino-4-(methylthio)buttersäure, (±)-Methionin, (±)-2-Amino-4-(methylmercapto)buttersäure

4030852.0025 4030852.0100
BACE4030852.0025EA 0 EUR
BACE4030852.0025 BACE4030852.0100
DL-Methionin
DL-Methionin
Formel: C₅H₁₁NO₂S
Molecular Weight: 149,21 g/mol
Schmelzpunkt: ∼136 °C
Dichte: 1,34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00063096
CAS-Nummer: 59-51-8
EINECS: 200-432-1
Merck Index: 13,06004

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR