(±)-3-Amino-1,2-propandiol 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: (±)-3-Amino-1,2-dihydroxy propane, 3-Amino-1,2-propanediol

B22672.14 B22672.22 B22672.36
B22672.14EA 67.7 EUR
B22672.14 B22672.22 B22672.36
(±)-3-Amino-1,2-propandiol 97%
(±)-3-Amino-1,2-propandiol

Purity: 97% CAS No.: 616-30-8 Molecular Formula: C3H9NO2 Formula Weight: 91.11 Flash Point: 186°(366°F) Refractive Index: 1.4920 Boiling Point: 264-266°/739mm Beilstein Reference: 1719121 UN No.: 2735 MDL No.: MFCD00008140

Formel: C₃H₉NO₂
Molecular Weight: 91,11 g/mol
Siedepunkt: 264…266 °C (739 mmHg)
Dichte: 1,186
Flammpunkt: 186 °C (366 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008140
CAS-Nummer: 616-30-8
EINECS: 210-475-8
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR