Benzylchlorid ≥99% stabilisiert
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Chlormethylbenzol, α-Chlortoluol, Chlorphenylmethan, (Chlormethyl)benzol
A12481.0E
A12481.36
A12481.0EEA
78.5
EUR
A12481.0E
A12481.36
Benzylchlorid ≥99% stabilisiert
Benzylchlorid
Stabilized
Purity: 99%
CAS No.: 100-44-7
Molecular Formula: C7H7Cl
Formula Weight: 126.59
Flash Point: 73°(163°F)
Physical Form: Liquid
Density: 1.099
Refractive Index: 1.5380
Melting Point: -43°
Boiling Point: 178-180°
Solubility: Miscible with alcohol, ether. Insoluble in water
Beilstein Reference: 471308
Merck Reference: 14,1129
UN No.: 1738
MDL No.: MFCD00000889
Formel:
(C₆H₅)CH₂Cl Siedepunkt: 178…180 °C Schmelzpunkt: –43 °C Dichte: 1,099 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000889 CAS-Nummer: 100-44-7 EINECS: 202-853-6 UN: 1738 ADR: 6.1,II Merck Index: 14,1129 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...