(±)-3-Methylvaleriansäure ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(±)-3-Methylpentansäure
A18886.06
A18886.14
A18886.22
A18886.06EA
33.9
EUR
A18886.06
A18886.14
A18886.22
(±)-3-Methylvaleriansäure ≥99%
(±)-3-Methylvaleriansäure
Purity: 99%
CAS No.: 105-43-1
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 85°(185°F)
Refractive Index: 1.4160
Odour: Unpleasant
Melting Point: -42°
Boiling Point: 196-198°
Beilstein Reference: 1720696
UN No.: 3265
MDL No.: MFCD00002729
Formel:
C₆H₁₂O₂ Molecular Weight: 116,16 g/mol Siedepunkt: 196…198 °C Schmelzpunkt: –42 °C Dichte: 0,930 Flammpunkt: 85 °C (185 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002729 CAS-Nummer: 105-43-1 EINECS: 203-297-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...