DL-Valin ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: (±)-2-Amino-3-methylbutansäure, H-DL-Val-OH, (±)-Valin, (±)-2-Aminoisovaleriansäure

A16756.0E A16756.22 A16756.36
A16756.0EEA 605 EUR
A16756.0E A16756.22 A16756.36
DL-Valin ≥99%
DL-Valin

Purity: 99% CAS No.: 516-06-3 Molecular Formula: C5H11NO2 Formula Weight: 117.15 Melting Point: ca 296° subl. Beilstein Reference: 506689 Merck Reference: 14,9909 MDL No.: MFCD00004267

Formel: C₅H₁₁NO₂
Molecular Weight: 117.15 g/mol
Siedepunkt: 217
Schmelzpunkt: ∼ 296 °C (Subl.)
Dichte: 1.352 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004267
CAS-Nummer: 516-06-3
EINECS: 208-220-0
Merck Index: 12,10046

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR