DL-Leucin ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: (±)-Leucin, DL-α-Amino-γ-methylvaleriansäure, (±)-α-Amino-γ-methylvaleriansäure, (±)-2-Amino-4-methylpentansäure

A10590.0B A10590.18 A10590.30
A10590.0BEA 641 EUR
A10590.0B A10590.18 A10590.30
DL-Leucin ≥99%
DL-Leucin

Purity: 99% CAS No.: 328-39-2 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: ca 294° subl. Beilstein Reference: 636005 Merck Reference: 14,5451 MDL No.: MFCD00063087

Formel: C₆H₁₃NO₂
Molecular Weight: 131,17 g/mol
Siedepunkt: 226 °C (1013 hPa)
Schmelzpunkt: ∼ 294 °C (Subl.)
Dichte: 1,293
Lagertemperatur: Raumtemperatur
MDL: MFCD00063087
CAS-Nummer: 328-39-2
EINECS: 206-328-2
Merck Index: 14,5451

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR