DL-Norvalin ≥98%
Lieferant: Thermo Scientific
Synonyme:
(±)-2-Aminovaleriansäure, (±)-Norvalin, H-DL-Nva-OH, (±)-2-Aminopentansäure
A15900.22
A15900.36
A15900.14
A15900.22EA
88.3
EUR
A15900.22
A15900.36
A15900.14
DL-Norvalin ≥98%
DL-Norvalin
Purity: 98%
CAS No.: 760-78-1
Molecular Formula: C5H11NO2
Formula Weight: 117.15
Melting Point:
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol Siedepunkt: 223 °C (1013 hPa) Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064420 CAS-Nummer: 760-78-1 EINECS: 212-082-7 Merck Index: 14,6716 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...