N-Acetyl-DL-Tryptophan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Ac-DL-Trp-OH, N(α)-Acetyl-DL-Tryptophan
M-Clarity™ Qualitätsstufe = MQ200
A6251-1KG
A6251-25G
A6251-5G
SIALA6251-1KGEA
1490
EUR
SIALA6251-1KG
SIALA6251-25G
SIALA6251-5G
N-Acetyl-DL-Tryptophan, Sigma-Aldrich®
N-Acetyl-DL-Tryptophan
Formel:
C₁₃H₁₄N₂O₃ Molecular Weight: 246,27 g/mol Schmelzpunkt: 205…206 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005644 CAS-Nummer: 87-32-1 EINECS: 201-739-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...