(±)-3-Phenylbuttersäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-3-Phenylbutansäure

M-Clarity™ Qualitätsstufe = MQ100

116807-25G 116807-5G
SIAL116807-25GEA 328 EUR
SIAL116807-25G SIAL116807-5G
(±)-3-Phenylbuttersäure, Sigma-Aldrich®
(±)-3-Phenylbuttersäure
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
MDL: MFCD00002725
CAS-Nummer: 4593-90-2
EINECS: 224-987-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR