DL-Tyrosin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyphenylalanin, 2-Amino-3-(4-hydroxyphenyl)propionsäure, H-DL-Tyr-OH, 3-(p-Hydroxyphenyl)-DL-alanin
A13740.09
A13740.18
A13740.30
A13740.09EA
35.6
EUR
A13740.09
A13740.18
A13740.30
DL-Tyrosin ≥98%
DL-Tyrosin
Purity: 98%
CAS No.: 556-03-6
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Melting Point: ca 325° dec.
Beilstein Reference: 515881
Merck Reference: 14,9839
MDL No.: MFCD00063074
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Siedepunkt: 378 °C (1013 hPa) Schmelzpunkt: ∼ 325 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063074 CAS-Nummer: 556-03-6 EINECS: 209-113-1 Merck Index: 14,9839 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...